Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK052340.1 | - | 6 - 7 | 991 | 61 | |
AK173233.1 | - | 3 - 4 | 380 | 61 | ||
BC059083.1 | - | 6 - 7 | 991 | 6827289 | 61 | |
BC062917.1 | - | 6 - 7 | 997 | 6831835 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |