Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sall4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK049188.1 | - | 1 - 2 | 219 | 104 | |
AK145388.1 | - | 1 - 2 | 237 | 104 | ||
AY463371.1 | - | 1 - 2 | 119 | 104 | ||
AY463373.1 | - | 1 - 2 | 119 | 104 | ||
BB867997.1 | - | 1 - 2 | 293 | 104 | ||
BC099928.1 | - | 1 - 2 | 242 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |