Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P201884 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P201884 |
AK009006.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P201884 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P201884 |
AK009006.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
159493 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
819291 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
42336 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693946 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
184651 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
220391 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
958006 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
746052 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743952 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
579061 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
129425 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
956835 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
859909 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
847893 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906990 |
AK003619.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906990 |
AK014633.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906990 |
BC106188.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906990 |
NM_025718.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
528619 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
672008 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491083 |
AK078072.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
622767 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
652557 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
452459 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
156864 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
559414 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
59193 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
436126 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
126785 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
453322 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
523201 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
970851 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
157803 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
106154 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
23754 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
51263 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
96249 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
176483 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P113214 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P111779 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P156644 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P115410 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P201884 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P201884 |
AK009006.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P201884 |
NM_023842.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P201884 |
AK009006.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P156644 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P113214 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P111779 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P115410 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P242544 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P242544 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P242544 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P549815 |
BC026631.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P549815 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
BC054853.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P101460 |
AK077574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P201884 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P201884 |
AK009006.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P201884 |
NM_023842.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P201884 |
AK009006.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115426 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P156644 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P111779 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P113214 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P115410 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959029 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958969 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959067 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10915926 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959079 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958981 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10656880 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958980 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959092 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10638409 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959046 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10638410 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959091 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11020691 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959040 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958996 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959083 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959043 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959048 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959041 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959055 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958993 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959103 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959024 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959071 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959109 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959006 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959032 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959090 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959026 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10971996 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959088 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958983 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958998 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959082 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959020 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959051 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959053 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10561792 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959012 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959107 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959072 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959074 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959108 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10605957 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959027 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959081 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959035 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959069 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959017 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959002 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959078 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10972007 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959076 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959110 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10972017 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959015 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959086 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10972013 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11017289 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10656882 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958985 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959004 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959105 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959025 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959060 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11003687 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958971 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959028 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10972005 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959097 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959102 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959106 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959009 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959034 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10591380 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959036 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10705548 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959037 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11077799 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959065 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10638419 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10860135 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959023 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10561789 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10972015 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10685113 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10779597 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959096 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10915902 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28959094 |
- |
- |