Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175419.4 | NP_780628.3 | 1 - 2 | 606 | 83 | |
GenBank mRNA | AK046312.1 | - | 1 - 2 | 632 | 83 | |
AK046584.1 | - | 1 - 2 | 598 | 83 | ||
AK142980.1 | - | 1 - 2 | 607 | 83 | ||
BC052039.1 | - | 1 - 2 | 587 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |