Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900017621.mm.1 |
NM_175481.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017621.mm.1 |
NM_175481.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017621.mm.1 |
TC0900002121.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017623.mm.1 |
NM_175481.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017623.mm.1 |
NM_175481.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017623.mm.1 |
TC0900002121.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017618.mm.1 |
NM_175481.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017618.mm.1 |
NM_175481.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017618.mm.1 |
TC0900002121.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017619.mm.1 |
NM_175481.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017619.mm.1 |
NM_175481.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017619.mm.1 |
TC0900002121.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017622.mm.1 |
NM_175481.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017622.mm.1 |
NM_175481.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017622.mm.1 |
TC0900002121.mm.1 |
MTA TC |