Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam76b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176836.3 | NP_789806.1 | 1 - 2 | 333 | 135 | |
GenBank mRNA | AK052430.1 | - | 1 - 2 | 291 | 135 | |
AK145026.1 | - | 1 - 2 | 345 | 135 | ||
BC043120.1 | - | 1 - 2 | 333 | 6408908 | 135 | |
BC050870.1 | - | 1 - 2 | 254 | 6335538 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |