Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177323.3 | NP_796297.2 | 3 - 4 | 371 | 151 | |
GenBank mRNA | AK028707.1 | - | 3 - 4 | 402 | 151 | |
AK036805.1 | - | 3 - 4 | 371 | 151 | ||
AK133789.1 | - | 3 - 4 | 397 | 151 | ||
AK135334.1 | - | 3 - 4 | 392 | 151 | ||
AK144144.1 | - | 3 - 4 | 393 | 151 | ||
BC065059.1 | - | 3 - 4 | 371 | 6401664 | 151 | |
BC145267.1 | - | 3 - 4 | 361 | 9053821 | 151 | |
BC150745.1 | - | 3 - 4 | 361 | 9087656 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |