Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177381.3 | NP_796355.2 | 10 - 11 | 1153 | 68 | |
GenBank mRNA | AK140362.1 | - | 10 - 11 | 1153 | 68 | |
BC038030.1 | - | 11 - 12 | 1164 | 4020725 | 68 | |
BC048005.1 | - | 6 - 7 | 508 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |