Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ofd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177429.3 | NP_803178.2 | 4 - 5 | 799 | 140 | |
GenBank mRNA | AJ438159.1 | - | 4 - 5 | 650 | 140 | |
BC119070.1 | - | 4 - 5 | 482 | 8733819 | 140 | |
BC120487.1 | - | 4 - 5 | 482 | 40129410 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |