Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177606.4 | NP_808274.2 | 10 - 11 | 1895 | 66 | |
GenBank mRNA | AK029252.1 | - | 10 - 11 | 1897 | 66 | |
AK173337.1 | - | 7 - 8 | 1551 | 66 | ||
BC125583.1 | - | 10 - 11 | 1860 | 40130098 | 66 | |
BC137804.1 | - | 10 - 11 | 1857 | 8860823 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |