Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177606.4 | NP_808274.2 | 15 - 16 | 2533 | 60 | |
GenBank mRNA | AK029252.1 | - | 15 - 16 | 2535 | 60 | |
AK173337.1 | - | 12 - 13 | 2188 | 60 | ||
BC125583.1 | - | 15 - 16 | 2498 | 40130098 | 60 | |
BC137804.1 | - | 15 - 16 | 2495 | 8860823 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |