Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177606.4 | NP_808274.2 | 4 - 5 | 410 | 122 | |
GenBank mRNA | AK029252.1 | - | 4 - 5 | 412 | 122 | |
AK173337.1 | - | 1 - 2 | 66 | 122 | ||
BC021518.1 | - | 4 - 5 | 382 | 122 | ||
BC125583.1 | - | 4 - 5 | 375 | 40130098 | 122 | |
BC137804.1 | - | 4 - 5 | 372 | 8860823 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |