Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000023067.mm.1 |
NM_001171026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023067.mm.1 |
NM_001171026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023067.mm.1 |
NM_177614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023067.mm.1 |
NM_177614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023067.mm.1 |
TC1000003097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000023069.mm.1 |
NM_001171026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023069.mm.1 |
NM_001171026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023069.mm.1 |
NM_177614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023069.mm.1 |
NM_177614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023069.mm.1 |
TC1000003097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000023066.mm.1 |
NM_001171026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023066.mm.1 |
NM_001171026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023066.mm.1 |
NM_177614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023066.mm.1 |
NM_177614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023066.mm.1 |
TC1000003097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000023068.mm.1 |
NM_001171026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023068.mm.1 |
NM_001171026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023068.mm.1 |
NM_177614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023068.mm.1 |
NM_177614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000023068.mm.1 |
TC1000003097.mm.1 |
MTA TC |