Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177231.2 | NP_796205.1 | 2 - 3 | 350 | 82 | |
NM_178220.3 | NP_835738.1 | 2 - 3 | 350 | 82 | ||
GenBank mRNA | AK052588.1 | - | 2 - 3 | 127 | 82 | |
AK090090.1 | - | 2 - 3 | 177 | 82 | ||
AK144054.1 | - | 2 - 3 | 149 | 82 | ||
AK147508.1 | - | 2 - 3 | 351 | 82 | ||
AK147566.1 | - | 2 - 3 | 371 | 82 | ||
AK165514.1 | - | 2 - 3 | 149 | 82 | ||
BC108969.1 | - | 2 - 3 | 117 | 40047224 | 82 | |
BC108970.1 | - | 2 - 3 | 117 | 40047225 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |