Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178381.3 | NP_848468.2 | 17 - 18 | 1548 | 71 | |
GenBank mRNA | BC025477.1 | - | 16 - 17 | 1595 | 71 | |
BC049815.1 | - | 7 - 8 | 1557 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |