Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200025619.mm.1 |
NM_001162485.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025619.mm.1 |
NM_001162485.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025619.mm.1 |
NM_178408.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025619.mm.1 |
NM_178408.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025619.mm.1 |
NR_027876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025619.mm.1 |
NR_027876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025619.mm.1 |
TC0200003122.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025618.mm.1 |
NM_001162485.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025618.mm.1 |
NM_001162485.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025618.mm.1 |
NM_178408.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025618.mm.1 |
NM_178408.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025618.mm.1 |
NR_027876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025618.mm.1 |
NR_027876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025618.mm.1 |
TC0200003122.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025622.mm.1 |
NM_001162485.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025622.mm.1 |
NM_001162485.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025622.mm.1 |
NM_178408.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025622.mm.1 |
NM_178408.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025622.mm.1 |
NR_027876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025622.mm.1 |
NR_027876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025622.mm.1 |
TC0200003122.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025616.mm.1 |
NM_001162485.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025616.mm.1 |
NM_001162485.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025616.mm.1 |
NM_178408.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025616.mm.1 |
NM_178408.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025616.mm.1 |
NR_027876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025616.mm.1 |
NR_027876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025616.mm.1 |
TC0200003122.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025617.mm.1 |
NM_001162485.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025617.mm.1 |
NM_001162485.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025617.mm.1 |
NM_178408.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025617.mm.1 |
NM_178408.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025617.mm.1 |
NR_027876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025617.mm.1 |
NR_027876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025617.mm.1 |
TC0200003122.mm.1 |
MTA TC |