Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178440.4 | NP_848534.2 | 15 - 16 | 2073 | 58 | |
GenBank mRNA | AK030825.1 | - | 15 - 16 | 2074 | 58 | |
AK036461.1 | - | 15 - 16 | 2058 | 58 | ||
AK042033.1 | - | 15 - 16 | 2025 | 58 | ||
AK088011.1 | - | 15 - 16 | 2074 | 58 | ||
AK146251.1 | - | 15 - 16 | 2046 | 58 | ||
AK152797.1 | - | 15 - 16 | 2067 | 58 | ||
AK159700.1 | - | 15 - 16 | 2049 | 58 | ||
AK171284.1 | - | 15 - 16 | 2067 | 58 | ||
AK171505.1 | - | 15 - 16 | 2047 | 58 | ||
BC028661.1 | - | 3 - 4 | 361 | 58 | ||
BC113755.1 | - | 15 - 16 | 2017 | 40061202 | 58 | |
BC113954.1 | - | 15 - 16 | 2017 | 40061200 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |