Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178609.4 | NP_848724.2 | 11 - 12 | 2341 | 68 | |
GenBank mRNA | AK031078.1 | - | 11 - 12 | 2339 | 68 | |
AK045040.1 | - | 11 - 12 | 2306 | 68 | ||
AK143686.1 | - | 11 - 12 | 2314 | 68 | ||
AY463359.1 | - | 11 - 12 | 2338 | 68 | ||
BC145429.1 | - | 11 - 12 | 2294 | 9053987 | 68 | |
BC150772.1 | - | 11 - 12 | 2294 | 9087683 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |