Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sccpdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178653.3 | NP_848768.1 | 6 - 7 | 820 | 84 | |
GenBank mRNA | AK042919.1 | - | 6 - 7 | 708 | 84 | |
AK046033.1 | - | 6 - 7 | 821 | 84 | ||
AK046468.1 | - | 6 - 7 | 808 | 84 | ||
AK050554.1 | - | 6 - 7 | 769 | 84 | ||
AK077695.1 | - | 6 - 7 | 770 | 84 | ||
AK082803.1 | - | 6 - 7 | 705 | 84 | ||
AK086448.1 | - | 6 - 7 | 843 | 84 | ||
AK086632.1 | - | 6 - 7 | 818 | 84 | ||
AK145399.1 | - | 6 - 7 | 804 | 84 | ||
AK166018.1 | - | 6 - 7 | 732 | 84 | ||
BC025803.1 | - | 6 - 7 | 817 | 5100552 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |