Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178662.3 | NP_848777.1 | 8 - 9 | 1206 | 71 | |
GenBank mRNA | AK028298.1 | - | 8 - 9 | 1213 | 71 | |
AK038483.1 | - | 8 - 9 | 1216 | 71 | ||
AK038990.1 | - | 8 - 9 | 1207 | 71 | ||
AK039411.1 | - | 8 - 9 | 1214 | 71 | ||
AK086723.1 | - | 8 - 9 | 1249 | 71 | ||
AK162959.1 | - | 8 - 9 | 1214 | 71 | ||
AY349150.1 | - | 8 - 9 | 1171 | 71 | ||
BC048903.1 | - | 8 - 9 | 1190 | 6491141 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |