Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178689.4 | NP_848804.2 | 5 - 6 | 842 | 62 | |
GenBank mRNA | AK032395.1 | - | 5 - 6 | 845 | 62 | |
BC023285.1 | - | 2 - 3 | 285 | 3984456 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |