Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3gap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178690.4 | NP_848805.2 | 4 - 5 | 325 | 137 | |
GenBank mRNA | AK076399.1 | - | 4 - 5 | 308 | 137 | |
AK080432.1 | - | 4 - 5 | 307 | 137 | ||
AK134604.1 | - | 4 - 5 | 307 | 137 | ||
AK164523.1 | - | 4 - 5 | 306 | 137 | ||
AK170768.1 | - | 4 - 5 | 297 | 137 | ||
AK172881.1 | - | 4 - 5 | 312 | 137 | ||
BC046297.1 | - | 4 - 5 | 291 | 6486170 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |