Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098476.2 | NP_001091946.1 | 8 - 9 | 1118 | 134 | |
NM_178700.5 | NP_848815.2 | 8 - 9 | 1446 | 134 | ||
GenBank mRNA | AK045986.1 | - | 8 - 9 | 1400 | 134 | |
AK077782.1 | - | 8 - 9 | 1447 | 134 | ||
AK089014.1 | - | 7 - 8 | 982 | 134 | ||
AK168385.1 | - | 8 - 9 | 1090 | 134 | ||
AK168459.1 | - | 8 - 9 | 1090 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |