Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plscr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178711.3 | NP_848826.1 | 7 - 8 | 1342 | 138 | |
GenBank mRNA | AK035919.1 | - | 7 - 8 | 1343 | 138 | |
AK045372.1 | - | 7 - 8 | 1344 | 138 | ||
AK054258.1 | - | 7 - 8 | 900 | 138 | ||
AK079413.1 | - | 2 - 3 | 308 | 138 | ||
AK082330.1 | - | 6 - 7 | 785 | 138 | ||
AK141580.1 | - | 868 | 138 | |||
AK165919.1 | - | 7 - 8 | 1343 | 138 | ||
AK166080.1 | - | 7 - 8 | 835 | 138 | ||
BC052067.1 | - | 7 - 8 | 896 | 5708712 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |