Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc51b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178933.2 | NP_849264.1 | 2 - 3 | 154 | 74 | |
GenBank mRNA | AK002463.1 | - | 2 - 3 | 285 | 74 | |
AK004188.1 | - | 2 - 3 | 337 | 74 | ||
AY279396.1 | - | 2 - 3 | 154 | 74 | ||
BC115606.1 | - | 1 - 2 | 133 | 40102690 | 74 | |
BC115607.1 | - | 1 - 2 | 133 | 40102691 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |