Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181403.2 | NP_852068.1 | 2 - 3 | 259 | 92 | |
GenBank mRNA | AK030717.1 | - | 1 - 2 | 92 | ||
AK150606.1 | - | 2 - 3 | 311 | 92 | ||
AK159309.1 | - | 2 - 3 | 332 | 92 | ||
AK165826.1 | - | 2 - 3 | 312 | 94 | ||
BC025865.1 | - | 2 - 3 | 259 | 5029143 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |