Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181410.3 | NP_852075.1 | 1 - 2 | 53 | 65 | |
GenBank mRNA | AK050227.1 | - | 1 - 2 | 37 | 65 | |
BC017515.1 | - | 1 - 2 | 30 | 4504077 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |