Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286040.1 | NP_001272969.1 | 15 - 16 | 2488 | 77 | |
NM_001286041.1 | NP_001272970.1 | 14 - 15 | 2425 | 77 | ||
NM_181413.4 | NP_852078.1 | 15 - 16 | 2488 | 77 | ||
GenBank mRNA | AK080878.1 | - | 4 - 5 | 1123 | 77 | |
AK129094.1 | - | 14 - 15 | 2359 | 77 | ||
AK147273.1 | - | 15 - 16 | 2423 | 77 | ||
AK160806.1 | - | 2 - 3 | 162 | 77 | ||
AK169828.1 | - | 4 - 5 | 434 | 77 | ||
BC010633.1 | - | 3 - 4 | 371 | 77 | ||
BC050847.1 | - | 15 - 16 | 2454 | 6507130 | 77 | |
BC057331.1 | - | 3 - 4 | 632 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |