Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supv3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181423.2 | NP_852088.1 | 2 - 3 | 362 | 96 | |
GenBank mRNA | AK031911.1 | - | 2 - 3 | 353 | 96 | |
AK143755.1 | - | 2 - 3 | 387 | 96 | ||
BC049796.1 | - | 2 - 3 | 362 | 6332552 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |