Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahcyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171000.2 | NP_001164471.2 | 9 - 10 | 1304 | 90 | |
NM_001171001.1 | NP_001164472.1 | 9 - 10 | 1024 | 90 | ||
NM_021414.6 | NP_067389.5 | 9 - 10 | 1307 | 90 | ||
GenBank mRNA | AK014539.1 | - | 9 - 10 | 1255 | 90 | |
AK053527.1 | - | 9 - 10 | 988 | 90 | ||
AK122382.1 | - | 7 - 8 | 1010 | 90 | ||
BC059255.1 | - | 5 - 6 | 479 | 90 | ||
BC079660.1 | - | 9 - 10 | 1217 | 6853550 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |