Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289524.1 | NP_001276453.1 | 1 - 2 | 198 | 72 | |
NM_024194.6 | NP_077156.2 | 1 - 2 | 198 | 72 | ||
GenBank mRNA | AK011726.1 | - | 1 - 2 | 179 | 72 | |
AK017814.1 | - | 1 - 2 | 167 | 72 | ||
AK034978.1 | - | 178 | 72 | |||
AK148748.1 | - | 1 - 2 | 157 | 72 | ||
AK163288.1 | - | 1 - 2 | 166 | 72 | ||
AK170052.1 | - | 1 - 2 | 178 | 72 | ||
CJ072347.1 | - | 1 - 2 | 201 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mus musculus 1700 Probe spans exons |