Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143776.1 | NP_001137248.1 | 5 - 6 | 610 | 75 | |
NM_001143777.1 | NP_001137249.1 | 6 - 7 | 714 | 75 | ||
NM_024244.4 | NP_077206.3 | 5 - 6 | 610 | 75 | ||
GenBank mRNA | AK004797.1 | - | 5 - 6 | 594 | 75 | |
AK032053.1 | - | 6 - 7 | 526 | 75 | ||
AK036299.1 | - | 6 - 7 | 677 | 75 | ||
AK041147.1 | - | 5 - 6 | 611 | 75 | ||
AK044049.1 | - | 6 - 7 | 637 | 75 | ||
AK082202.1 | - | 5 - 6 | 606 | 75 | ||
AK220421.1 | - | 6 - 7 | 550 | 75 | ||
BC129839.1 | - | 5 - 6 | 566 | 40060810 | 75 | |
BC129840.1 | - | 5 - 6 | 566 | 40060811 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |