Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110015.1 | NP_001103485.1 | 12 - 13 | 1504 | 84 | |
NM_001110016.1 | NP_001103486.1 | 12 - 13 | 1504 | 84 | ||
NM_144863.4 | NP_659112.2 | 12 - 13 | 1504 | 84 | ||
GenBank mRNA | AK041763.1 | - | 12 - 13 | 1424 | 84 | |
AK045396.1 | - | 12 - 13 | 1477 | 84 | ||
AK077545.1 | - | 12 - 13 | 1433 | 84 | ||
AK155965.1 | - | 12 - 13 | 1402 | 84 | ||
AK171683.1 | - | 12 - 13 | 1442 | 84 | ||
AK172054.1 | - | 12 - 13 | 1429 | 84 | ||
AK172377.1 | - | 12 - 13 | 1434 | 84 | ||
AK186642.1 | - | 1 - 2 | 99 | 84 | ||
AK214226.1 | - | 1 - 2 | 99 | 84 | ||
BC022132.1 | - | 12 - 13 | 1476 | 84 | ||
BC025809.1 | - | 12 - 13 | 1418 | 84 | ||
BC094453.1 | - | 12 - 13 | 1455 | 5068941 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |