Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175124.5 | NP_780333.3 | 6 - 7 | 713 | 92 | |
NR_028143.1 | 6 - 7 | 618 | 92 | |||
NR_028144.1 | 5 - 6 | 577 | 92 | |||
GenBank mRNA | AK004904.1 | - | 6 - 7 | 587 | 92 | |
AK077270.1 | - | 6 - 7 | 709 | 92 | ||
AK159417.1 | - | 6 - 7 | 682 | 92 | ||
BC027058.1 | - | 3 - 4 | 270 | 92 | ||
BC052051.1 | - | 4 - 5 | 484 | 92 | ||
BC086694.1 | - | 6 - 7 | 604 | 30697866 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |