Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175155.4 | NP_780364.3 | 15 - 16 | 2134 | 88 | |
GenBank mRNA | AJ507736.1 | - | 15 - 16 | 1949 | 88 | |
AK039638.1 | - | 3 - 4 | 473 | 88 | ||
AK129216.1 | - | 10 - 11 | 1468 | 88 | ||
AK149872.1 | - | 15 - 16 | 2086 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |