Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100009571.mm.1 |
NM_001145804.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009571.mm.1 |
NM_001145804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009571.mm.1 |
NM_175294.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009571.mm.1 |
NM_175294.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009571.mm.1 |
TC0100001178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009573.mm.1 |
NM_001145804.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009573.mm.1 |
NM_001145804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009573.mm.1 |
NM_175294.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009573.mm.1 |
NM_175294.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009573.mm.1 |
TC0100001178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009574.mm.1 |
NM_001145804.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009574.mm.1 |
NM_001145804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009574.mm.1 |
NM_175294.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009574.mm.1 |
NM_175294.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009574.mm.1 |
TC0100001178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009572.mm.1 |
NM_001145804.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009572.mm.1 |
NM_001145804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009572.mm.1 |
NM_175294.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009572.mm.1 |
NM_175294.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009572.mm.1 |
TC0100001178.mm.1 |
MTA TC |