Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030180.2 | NP_084456.2 | 7 - 8 | 1421 | 82 | |
GenBank mRNA | AK036864.1 | - | 6 - 7 | 810 | 82 | |
AK047620.1 | - | 7 - 8 | 1421 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |