Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177045.3 | NP_796019.1 | 9 - 10 | 1147 | 105 | |
GenBank mRNA | AK043839.1 | - | 9 - 10 | 1148 | 105 | |
AK146601.1 | - | 9 - 10 | 1166 | 105 | ||
AK173271.1 | - | 5 - 6 | 575 | 105 | ||
BC055068.1 | - | 7 - 8 | 789 | 105 | ||
BC062883.1 | - | 2 - 3 | 185 | 105 | ||
BC125517.1 | - | 8 - 9 | 1037 | 40130032 | 105 | |
BC137743.1 | - | 8 - 9 | 1037 | 8860762 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |