Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900007034.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007034.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007034.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007037.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007037.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007037.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007040.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007040.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007040.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007041.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007041.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007041.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007026.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007026.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007026.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007032.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007032.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007032.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007042.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007042.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007042.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007029.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007029.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007029.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007030.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007030.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007030.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007043.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007043.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007043.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007028.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007028.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007028.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007031.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007031.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007031.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007038.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007038.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007038.mm.1 |
TC0900000917.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007039.mm.1 |
NM_145618.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007039.mm.1 |
NM_145618.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007039.mm.1 |
TC0900000917.mm.1 |
MTA TC |