Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177152.5 | NP_796126.4 | 6 - 7 | 1058 | 94 | |
GenBank mRNA | AK035166.1 | - | 6 - 7 | 1060 | 94 | |
AK137007.1 | - | 6 - 7 | 1045 | 94 | ||
AY505341.1 | - | 6 - 7 | 1001 | 94 | ||
BC065142.1 | - | 6 - 7 | 1024 | 30610561 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |