Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4ha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177161.4 | NP_796135.3 | 3 - 4 | 574 | 60 | |
GenBank mRNA | AK086468.1 | - | 1 - 2 | 162 | 60 | |
AK086780.1 | - | 3 - 4 | 556 | 60 | ||
AY313449.1 | - | 3 - 4 | 574 | 60 | ||
BC082538.1 | - | 3 - 4 | 616 | 5701368 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |