Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037740.1 | NP_001032829.1 | 6 - 7 | 694 | 82 | |
NM_177204.3 | NP_796178.2 | 6 - 7 | 694 | 82 | ||
NR_104576.1 | 6 - 7 | 694 | 82 | |||
GenBank mRNA | AK041004.1 | - | 6 - 7 | 695 | 82 | |
AK042073.1 | - | 6 - 7 | 695 | 82 | ||
AK052271.1 | - | 6 - 7 | 695 | 82 | ||
AK122459.1 | - | 6 - 7 | 499 | 82 | ||
AK163706.1 | - | 6 - 7 | 694 | 82 | ||
BC147502.1 | - | 6 - 7 | 686 | 9056385 | 82 | |
BC172034.1 | - | 6 - 7 | 686 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |