Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177213.3 | NP_796187.2 | 4 - 5 | 618 | 102 | |
GenBank mRNA | AK029712.1 | - | 4 - 5 | 618 | 102 | |
AY243471.1 | - | 4 - 5 | 618 | 102 | ||
BC141350.1 | - | 4 - 5 | 583 | 9055962 | 102 | |
BC141351.1 | - | 4 - 5 | 583 | 9055963 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |