Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205227.1 | NP_001192156.1 | 1 - 2 | 80 | 124 | |
NM_177216.4 | NP_796190.1 | 1 - 2 | 80 | 124 | ||
GenBank mRNA | AK085272.1 | - | 1 - 2 | 45 | 124 | |
AK147952.1 | - | 1 - 2 | 81 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |