Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177236.4 | NP_796210.2 | 5 - 6 | 1158 | 67 | |
GenBank mRNA | AK032322.1 | - | 5 - 6 | 1083 | 67 | |
AK045516.1 | - | 5 - 6 | 1158 | 67 | ||
AK047008.1 | - | 2 - 3 | 4063 | 67 | ||
BC118975.1 | - | 4 - 5 | 890 | 40098391 | 67 | |
BC118976.1 | - | 4 - 5 | 890 | 40098398 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |