Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146043.2 | NP_001139515.1 | 4 - 5 | 318 | 100 | |
NM_001271463.1 | NP_001258392.1 | 4 - 5 | 318 | 100 | ||
NM_001271464.1 | NP_001258393.1 | 4 - 5 | 318 | 100 | ||
NM_001271465.1 | NP_001258394.1 | 4 - 5 | 345 | 100 | ||
NM_177266.5 | NP_796240.3 | 4 - 5 | 351 | 100 | ||
GenBank mRNA | AK028450.1 | - | 4 - 5 | 304 | 100 | |
AK035819.1 | - | 4 - 5 | 257 | 100 | ||
AK044371.1 | - | 4 - 5 | 319 | 100 | ||
AK083441.1 | - | 4 - 5 | 100 | |||
AK161797.1 | - | 4 - 5 | 254 | 100 | ||
AK165871.1 | - | 4 - 5 | 312 | 100 | ||
AK219489.1 | - | 3 - 4 | 164 | 100 | ||
BB841178.1 | - | 4 - 5 | 312 | 100 | ||
BC027341.1 | - | 4 - 5 | 302 | 3984129 | 100 | |
BC138233.1 | - | 4 - 5 | 249 | 8861252 | 100 | |
BC145208.1 | - | 4 - 5 | 249 | 9053759 | 100 | |
BY742160.1 | - | 4 - 5 | 319 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |