Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159389.1 | NP_001152861.1 | 8 - 9 | 906 | 66 | |
NM_177306.3 | NP_796280.1 | 5 - 6 | 537 | 66 | ||
GenBank mRNA | AK029809.1 | - | 5 - 6 | 538 | 66 | |
AK049380.1 | - | 8 - 9 | 886 | 66 | ||
BC132104.1 | - | 3 - 4 | 220 | 40130381 | 66 | |
BC132294.1 | - | 3 - 4 | 220 | 40130571 | 66 | |
BC145065.1 | - | 3 - 4 | 220 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |