Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177693.3 | NP_808361.1 | 3 - 4 | 384 | 79 | |
GenBank mRNA | AK029354.1 | - | 3 - 4 | 385 | 79 | |
AK145594.1 | - | 1493 | 79 | |||
AK164958.1 | - | 3 - 4 | 383 | 79 | ||
BC119253.1 | - | 3 - 4 | 365 | 8734002 | 79 | |
BC119255.1 | - | 3 - 4 | 365 | 8734004 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |