Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700008257.mm.1 |
TC0700000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700008257.mm.1 |
NM_001271879.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008257.mm.1 |
NM_177694.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008257.mm.1 |
NR_073508.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008256.mm.1 |
TC0700000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700008256.mm.1 |
NM_001271879.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008256.mm.1 |
NM_177694.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008256.mm.1 |
NR_073508.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008255.mm.1 |
TC0700000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700008255.mm.1 |
NM_001271879.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008255.mm.1 |
NM_177694.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008255.mm.1 |
NR_073508.1 |
Refseq |