Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177697.3 | NP_808365.2 | 25 - 26 | 2948 | 63 | |
GenBank mRNA | AK086942.1 | - | 24 - 25 | 2546 | 63 | |
AK136898.1 | - | 24 - 25 | 2547 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |